Binocular magnifiers
Leverandør: ESCHENBACH OPTIK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
164451 164452 164453
630-8214EA 2590 DKK
630-8214 630-8215 630-8216
Binocular magnifiers
Forstørrelsesglas og lupper
Magnifiers with discrete metal frame and black matt finish for use with monocular lens components.
* Shorter working distance due to integrated convergence supporting prisms.
Leveringsinformation: Supplied with a protective box.
Advarsel: Not suitable for spectacle wearers.