UV-lamper og UV-kabinet
Leverandør: CAMAG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
022.9055 022.9060 022.9160 040.2000 040.2100
552-0050EA 0 DKK
552-0050 554-6593 554-6592 CAMA040.2000 CAMA040.2100
UV-lamper og UV-kabinet
Lyskilder UV-lamper
UV-lamper med stødsikkert kabinet, designet primært til TLC-laboratorier.
  • User friendly and versatile
  • Available in two wavelengths: 254/366 nm
  • Automatic switch off timer after 10 minutes reduces the risk of exposure to radiation
  • UV lamp 4: Tilt sensor automatically turns off the lamp in case the lamp is tilted more than 30 degrees
The viewing box is suitable for inspecting thin layer chromatograms and other objects in an undarkened room. The front of the box is closed with a roller shutter which can be pushed open to the left or to the right as required for inserting or marking objects. A glass filter in the viewing window protects the eyes against reflected short wave UV light.
UV Cabinet 4, a combination of the UV Lamp 4 and the Viewing Box 4, is specially designed for UV observation with minimal influence of ambient light. Thanks to a reduced footprint, UV Cabinet 4 requires only minimum space. The redesigned observation port has a built-in UV filter in the viewing window ensuring effective eye protection. The interior is accessible via the established roller shutter on the front.
Leveringsinformation: UV lamps are supplied as 2×8 W or 1×8 W light tubes. UV cabinet includes viewing box and lamps. Viewing boxes sold separately do not include lamps.