Håndholdt UV lampe
Leverandør: UVP ULTRA VIOLET PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
95-0104-02 95-0006-03 95-0004-10 95-0007-06 95-0005-06
730-1024EA 6630 DKK
730-1024 733-2421 732-1109 732-0857 733-2365
Håndholdt UV lampe
Lyskilder UV-lamper
These lightweight lamps can be used with the optional lamp stand for hands-free operation. Replacement tubes are also available.
  • Lightweight for easy handling
  • Durable plastic housing
  • Six watts
  • Models available with short wave, long wave UV or a combination short wave/long wave version
  • Combination short wave/long wave version comes with a snap-on wavelength selector for switching between wavelengths (UVGL-58 only)
W×D×H: 81×378×64 mm.
Weight: 0,9 kg.