Ice machine, cubes
Leverandør: PORKKA SCANDINAVIA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
KL22 KL32 KL42
474-0093EA 11670 DKK
474-0093 474-0094 474-0095
Ice machine, cubes
Ismaskiner
Ice machine in stainless steel for the production of ice cubes. Capacity from 18 to 145 kg/24 h. The ice machines are delivered with air cooling as a standard but are also available with water cooling. Water cooling is recommended for integrated machinery. Models with higher capacity are available. Please contact your local Avantor sales office for further details.