Gas burner, powerjet/powerjet 2
Leverandør: WARTEWIG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6.005.000 6.000.855 6.006.000
451-0112EA 2200 DKK
451-0112 451-0125 451-0126
Gas burner, powerjet/powerjet 2
Varmeapparater Gasbrændere
Mobile handheld laboratory gas burner with piezo ignition. The solution for flame sterilisation in the microbiological or biotechnological fields. The burner is lit as soon as the bar is pressed. The flame can be regulated in size and is extinguished by release of the bar.
  • Equipped with a locking device comprising both short and long term ranges
  • Powerjet for operation with a gas cartridge (eg. CG 1750 by Camping Gaz)
  • Powerjet 2 can be connected to a central gas supply or can be operated by gas cartridges (eg. CV360 from Camping Gaz)