Bunsen burners
Leverandør: USBECK Laborgeräte
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1000 1001 1010 1011 1030 1031 1050 1010/1 1011/1
451-0075EA 386 DKK
451-0075 451-0076 451-0077 451-0078 451-0079 451-0080 451-0083 451-1367 451-1368
Bunsen burners
Varmeapparater Gasbrændere
Nickel plated brass with cast iron foot.
  • With air regulation ring
Height: 145 mm (451-0079, 451-0080: 150 mm, 451-0083: 160 mm)
Head Ø: 17 mm (14 mm for propane models)