Teclu burner, with needle valve, 1300°C
Leverandør: BOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7400 7410
451-0006EA 1860 DKK
451-0006 451-0007
Teclu burner, with needle valve, 1300°C
Varmeapparater Gasbrændere
DIN 30665 version, nickel-plated brass.
  • Needle valve
  • With pilot light and air regulator
Burner Ø: 13 mm
Baseplate Ø: 76 mm
External tubing Ø: 10 mm
Height: 180 mm