Micro burners with needle valve, 1000 °C
Leverandør: BOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7300 7310
451-0001EA 685 DKK
451-0001 451-0002
Micro burners with needle valve, 1000 °C
Varmeapparater Gasbrændere
Nickelled brass.
  • Max. temperature 1000 °C
  • With air regulation
  • Accepts 10 mm Ø tubing
Height: 90 mm
Head Ø: 8 mm