Gasflaskeskab, type 30, G-CLASSIC-30
Leverandør: asecos
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30651001 30663001 30673001 30685001 30686001
135-1826EA 29230 DKK
135-1826 135-1829 135-1832 135-1835 135-1837
Gasflaskeskab, type 30, G-CLASSIC-30
Møbler Opbevaringskabinetter
Fire-resistant type 30 safety cabinets for the safe storage of gas cylinders in buildings, in accordance with DIN EN 14470-2.
  • Easy installation of equipment and pipes - large interior height (1890 mm) and adjusting aids to compensate for uneven floor
  • Access possibilities on the top of the cabinet
  • No unauthorised use - doors lockable with cylinder lock (suitable for locking systems)
  • Door opening angle max. 180°
  • Quickly put into operation - integrated air ducts ready for connection to a technical exhaust system, complete and uniform ventilation of the cabinet interior from the bottom to the top
30 minutes fire rated
Furnace tested (type tested)
GS approval, CE conformal
Labelling in accordance with EN 14470-2
Certificeringer: 30 minutes fire rated
Furnace tested (type tested)
GS approval, CE conformal
Labelling in accordance with EN 14470-2
Leveringsinformation: Connection to ventilation not included (available on request). Interior fittings must be ordered separately.
asecos equipment is delivered on a skid/pallet as standard. However, in view of the weight and relative fragility of this product (impact may damage the entire cabinet irreparably if the walls are cracked), it is advisable to select the 'delivery to installation site' service, which is delivery to the exact location in the laboratory where the cabinet is to be used. Based on the checklist (to be confirmed at the time the quotation is prepared) and the selection of the option of 'delivery to installation site', specialist delivery personnel will use custom handling equipment to install the equipment correctly. 'Delivery to installation site' will ensure the guaranteed conditions of the equipment are met. Delivery prices available on request.