Well visualisation plates
Leverandør: EMBITEC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EP-1047 EP-1048
730-2202EA 139 DKK
730-2202 730-2203
Well visualisation plates
Elektroforesesystemer Horisontale Gelelektroforesesystemer
RunOne™ well visualisation plates are designed to make gel loading simple. The light colour of most gel units provides little or no contrast for the agarose gel and the gel wells. RunOne™ well visualisation plates are red, fit securely into the RunOne™ system, and provide a stable platform for gel running.