Dissolved oxygen sensors for industrial applications, OxyFerm FDA
Leverandør: HAMILTON BONADUZ
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
237453 237454 237450 237452 237455 237540 237541 237542 237543 237544
662-0175EA 19600 DKK
662-0175 664-0202 664-0200 664-0201 664-0203 664-0222 664-0221 664-0220 664-0219 664-0218
Dissolved oxygen sensors for industrial applications, OxyFerm FDA
Elektroder Elektroder til opløst ilt
These electrochemical oxygen sensors are ideal for applications with high demands for hygiene, e.g. in pharmaceutical industry, in biotechnology and in foods & beverages production. The sensors are equipped with a FDA approved membrane for usage in hygienic processes. They can withstand steam sterilisation, autoclavation and CIP cleanings.
  • Steam sterilisation, cleaning in place (CIP), autoclavable
  • Low drift for reliable measurements and fast response
  • Reduced cost of ownership due to prolonged service cycles
Technical specifications
O₂: 10 ppb to saturation or 0,1 to 200% of air.
Temperature: 0 to 130 °C; max. 4 bar.
TC: 22 KOhm NTC.
ATEX / IECEx approval: CE 0035 ll 1/2 G Ex ia llC T4/T5/T6 Ga/Gb; CE 0035 ll 1/2 D ia lllC T x °C Da/Db
Certificeringer: ATEX/IECEx approval: CE 0035 ll 1/2 G Ex ia llC T4/T5/T6 Ga/Gb; CE 0035 ll 1/2 D ia lllC T x °C Da/Db.