Arsenic determination apparatus, Quickfit®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AD3 1AD/1
BIBBAD3EA 1950 DKK
BIBBAD3 201-8003
Arsenic determination apparatus, Quickfit®
Destruktionssystemer Apparatur til Detektion af Sporelementer
PYREX®, borosilicate glass arsenic determination apparatus designed for the estimation of arsenic and antimony.
  • Method uses the formation of a complex between arsine and silver diethyldithiocarbamate with subsequent measurement at 5380A
  • Complete assembly (all parts single available)