Calibration thermostats, RE and Proline range
Leverandør: LAUDA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
L001919 L001920 L001921 L001922 L001923 L001924 L001925 L001926 L001927 L001928 L001929 L001930 L001931 L001932 L001933 L001934
LAUDL001919EA 70520 DKK
LAUDL001919 LAUDL001920 LAUDL001921 LAUDL001922 461-1202 461-1203 461-1200 461-1201 LAUDL001927 LAUDL001928 LAUDL001929 LAUDL001930 LAUDL001931 LAUDL001932 LAUDL001933 LAUDL001934
Calibration thermostats, RE and Proline range
Cirkulatorsystemer Åbne cirkulatorsystemer Kalibreringstermostater
Calibration thermostats have been designed for applications in calibration laboratories such as ISO, DKD, NAMAS, UKAS and conform to the requirements according to DIN EN ISO 9001:2000. Units offer excellent temperature stability and distribution over a wide temperature range in the test chamber. They have a height adjustable usable space and a separate pre-chamber. All models have an RS232 interface. Connection for PT100 reference sensor except RE 212 J.
  • Temperature uniformity of ±0,01 K
  • RE models have a vario pump with 5 output steps, PJL units have varioflex pressure pump with 8 selectable output steps and UB models have a pressure pump
  • RE models can be used at temperatures below ambient, RE 212 J has the E 200 controller, and RE 312 J has the more sophisticated E 300 controller
  • PJL and UB -JL models can be used with the DLK 45 flow through cooler down to −40 °C
The overflow principle provides excellent temperature uniformity, the pump moves the bath liquid to the usable space. The level overflow ensures that the level is constant in the testing chamber. As the liquid level in the usable space can be changed a variety of test objects can be immersed and calibrated in accordance with the relevant standards.
Maximum pump capacities:
RE-J 17 L/min, 400 mbar pressure
PJ or PJL 25 L/min, 800 mbar pressure
Advarsel: * Cooling capacity at +20 °C
** with an external cooling source
*** the unit is 56 mm higher with the Command console.