VWR®, Fjerntemperaturfølere til VWR termostatbade
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
461-0101EA 3070 DKK
461-0101 461-0102 461-0103
VWR®, Fjerntemperaturfølere til VWR termostatbade
Temperatur prober
Remote temperature probes for circulator use with a chiller. Allows points of control to be switched from inside a chiller to a remote location. Compensates for heat loss through tubing between the circulator and the control point. Platinum RTD sensor with stainless steel sheath. Rubber coupling with 25,4 mm Ø, connects sheath to flexible cable. Probe cable ends in a DB9 female plug.
Længdeר: 152×4 mm