Temperaturtabelskrivere, SK4 og SL4
Leverandør: DICKSON
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SK4350 SL4100 SL4350
620-1502EA 0 DKK
620-1502 620-1503 620-1504
Temperaturtabelskrivere, SK4 og SL4
Skrivere Skrivere
These self-contained temperature recorders on a Ø 100 mm disc provide a permanent record of temperatures with an average response time of 11 minutes to 63% of full scale. Powered by a AA battery, they are used for monitoring and preparing stocks of vaccines, foods and pharmaceutical products. SL4 models include an LCD screen for displaying data (W×D: 48×16 mm).
  • Long term recording during storage, transport or shipping
  • Hinged door
  • Commuter for recording 7 days or 24 hours
W×D×H: 132×132×61 mm
Weight: 295 g