Rotors for Centrifuges 5804(R) and 5810(R)
Leverandør: EPPENDORF
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5804709.004 5804715.004 5804719.000 5804726.006 5804727.002 5804730.003 5804740.009 5804735.005 5820715.006 5804746.007 5820771.003 5820772.000 5820760.001 5820765.003
521-0044EA 11400 DKK
521-0044 521-0046 522-2454 521-0049 521-0050 521-0051 521-0055 521-0054 521-2073 521-1957 521-2080 521-2081 521-2076 521-2078
Rotors for Centrifuges 5804(R) and 5810(R)
Centrifuger Rotorer
Eppendorf rotors for centrifuges 5804(R) and 5810(R) precision meets power. Customise your centrifugation with varied capacities and speeds. Accessorise with versatile buckets and adaptors for efficient sample processing. Ideal for high-performance applications, ensuring accuracy and reproducibility.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 min)
  • Accessories (adaptors and buckets) further customise your centrifuge setup
  • Aerosol-tight options available
Experience precision in high-performance centrifugation with Eppendorf Rotors designed for centrifuge 5804/5804 R. These rotors offer diverse capacities and speeds, providing flexibility for various sample processing needs. Accessorise with versatile buckets and adaptors, enhancing the adaptability of your centrifuge setup. Ideal for high-performance applications, the centrifuge 5804 series is further optimised with these precision-engineered rotors, ensuring accuracy and reproducibility in every spin. Explore the range to elevate your centrifugation experience.
Bestillingsoplysninger: Rotors, adapters, and accessories are only for use with centrifuge models specified. Unless otherwise specified, rotors may be used with all centrifuge models.