VWR®, Modular Heating Blocks and Accessories for Block Heaters
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
460-3211EA 1080 DKK
460-3211 460-3235 460-3212 460-3234 460-3223 460-3221 460-3245 460-3246
VWR®, Modular Heating Blocks and Accessories for Block Heaters
Bloktermostater Blokke
Constructed from a solid anodised aluminium block.
  • Close contact of tubes to block walls allows for maximum heat transfer
  • Each block has a thermometer well for measuring block temperature
Dimensioner for enkeltblok (B×D×H): 97×75×51 mm
Dobbeltblok W×D×H: 95×152×57 mm