Mini Coolers
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
114930 114935 114940
479-1241EA 1250 DKK
479-1241 479-1242 479-1243
Mini Coolers
Bokse Kølere/Frysebokse/Poser
PC, yellow, with clear lid. Maintains temperature of 0 °C for 60 min.
  • Durable polycarbonate filled with non-toxic gel
  • Mini coolers hold twelve 0,5 to 2,0 ml tubes
Mini coolers are designed to protect a wide range of solutions (enzymes, DNA, RNA, cell suspensions) by helping to maintain freezer temperatures on the lab bench.