Cent-O-Gram® and Dial-O-Gram®
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
80000009 80000010
611-5003EA 3630 DKK
611-5003 611-5068
Cent-O-Gram® and Dial-O-Gram®
Vægte
Removable stainless steel pan, aluminium pressure cast base and beam. These balances offer high precision and excellent value and are globally popular in education and industry.
  • Self-aligning beam design
  • Magnetic dampening minimises oscillation and speeds weighing
  • Pan Ø 89 and 7 mm deep
  • Quick-zeroing with counterbalancing knob on Dial-O-Gram