Håndklæder, foldede og på rulle, Tork
Leverandør: Essity
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
290016 290076 100297 290067 100288 120181 290068 100278 290163 290179 290190 290059 150299
115-0225EA 1080 DKK
115-0225 115-0325 115-3108 115-1398 115-1575 115-1587 115-1363 115-1574 115-1726 115-1439 115-1634 115-1937 115-1941
Håndklæder, foldede og på rulle, Tork
Papirservietter Engangsservietter og - håndklæder Papirhåndklæder
Interfold, ZZ-and C-fold hand towels and hand towel rolls for Tork dispenser systems, various quality grades available.
  • Tork Premium: Soft premium quality for heaviest wiping tasks - with a better and faster absorption than other towels and wipes
  • Tork Advanced: Improved performance tissues and wipes that meet or exceed basic requirements - with a good absorption, brightness and strength
  • Tork Universal: Standard quality tissues and wipes for basic demands
Hand towel rolls for H1 dispenser
- Large number of towels on each roll: Less frequent refilling needed
- Highly absorbent: Towel dries quickly and thoroughly
Interfold hand towels for H2 dispenser
- Reduces consumption: Only one towel at a time
- Provides quick and effective hand drying
- Highly absorbent: Towel dries quickly and thoroughly
ZZ-fold and C-fold hand towels for H3 dispenser
- Traditional hand towel system with towels of short length
- C-fold enables user to take more than one towel at a time