Klude, formættede, TechniSat®
Leverandør: Texwipe
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TX1041 TX1065
115-0298EA 7020 DKK
115-0298 TEXWTX1065
Klude, formættede, TechniSat®
Papirservietter Servietter til Renrum Imprægneret
These pre-wetted wipes are made of a fabric blend of 55% non woven hydroentangled cellulose and 45% polyester.
  • Convenient wipes offer portion control and reduced potential of spillage in and out of the cleanroom
  • Excellent bi-directional strength and a very low extractable level
  • Ideal for cleaning environmental cleanroom surfaces during and following the production flow
  • Suitable for wiping of production equipment requiring low solvent extractables and low particle generation
This fabric combines the highly absorbent properties of a natural fibre with the cleanliness and strength of a synthetic fibre. It offers low particle counts, low extractable levels and solvent compatibility. TechniSat® wipes are pre-wetted with isopropyl alcohol/deionised water solutions filtered to 0,2 μm. The wipes are packed in resealable FlexPacks, canisters or slider bags.
Farve: Hvid
Suitable for use in Class 100 - 100000, ISO 5 - 8 cleanrooms.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904