VWR® ND11, krympehalshætteglas
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
548-0003EA 0 DKK
548-0003 548-0004 548-0005 548-0447 548-0906 548-2126
VWR® ND11, krympehalshætteglas
Hætteglas
These vials are for use on instruments from the following manufacturers: Agilent, Carlo Erba, CTC, DANI, Fisons, Gerstel, Jasco, PerkinElmer, Shimadzu, Spark, Thermo Scientific, Varian, etc.
  • Standard vials for GC and HPLC
  • Vials with a barcode label, pre-crimped vials are also available
  • Microlitre vials for sample preparation (reactions, concentrations) or as an alternative for conical micro-vials or crimp neck vials with inserts
TopSert vials are an alternative to glass vials with fused-in micro-inserts or micro-insert with plastic spring. The TopSert glass micro-insert is centred in the plastic mould and pressed firmly against the septa due to its slightly overlapping edge.
Leveringsinformation: Kits include: 1,5 ml glass crimp neck vial (11,6×32 mm), aluminium crimp cap, clear lacquered with 5,5 mm central hole and septum.