Sample bottles with or without push-on cap
Leverandør: GRAVIS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
E00910M E00914M E00936M E00915M E01020-100 E01144-68 E00925M E01025M E00920 E00920M E00910
215-2623EA 899 DKK
215-2623 215-2399 215-2630 215-2625 215-2632 SPVAE01144-68 215-3233 215-2409 SPVAE00920 SPVAE00920M SPVAE00910
Sample bottles with or without push-on cap
Flasker Glasflasker
Chemistry glass, white or amber, with or without natural PE push-on cap, assembled.
  • Amber glass to protect light-sensitive materials.
Bestillingsoplysninger: Replacement PE caps are available separately.