Dialysemembran Spectra/Por® 3
Leverandør: Spectrum Laboratories
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
132720 132724 132725
734-0684EA 4660 DKK
734-0684 734-0686 734-0687
Dialysemembran Spectra/Por® 3
Slanger Dialyseslanger
MWCO 3,5 kD, regenereret cellulose.
  • Transparent og fleksibel
This economical membrane is ideal for a broad range of applications in which there is a significant size difference between the MW species being separated; including sample prep, desalting, buffer exchange, pH change, protein purification. Offering a good chemical compatibility, Standard Grade RC can be used with dilute strong acids and bases, concentrated weak acids and bases, most alcohols and some mild or dilute organics, including DMSO. Standard RC can tolerate pH 2 to 12 and temperatures 4 to 121 °C. Supplied dry, preservative: glycerol.