Dialysemembran Spectra/Por® 2
Leverandør: Spectrum Laboratories
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
132676 132678 132680 132682 132684
734-0671EA 5250 DKK
734-0671 734-0672 734-0673 734-0675 734-0676
Dialysemembran Spectra/Por® 2
Slanger Dialyseslanger
MWCO 12 to 14 kD, regenerated cellulose.
  • Transparent and flexible
This economical membrane is ideal for a broad range of applications in which there is a significant size difference between the MW species being separated; including sample prep, desalting, buffer exchange, pH change, protein purification. Offering a good chemical compatibility, Standard Grade RC can be used with dilute strong acids and bases, concentrated weak acids and bases, most alcohols and some mild or dilute organics, including DMSO. Standard RC can tolerate pH 2 to 12 andMore Product Information