Tubing, PTFE
Leverandør: DEUTSCH NEUMANN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3400204 3402732 3400315 3400102 3400103 3400115 3400203 3400305 3400405 3400501 3400506 3400516 3400607 3400613 3400810 3400813 3401012 3401214 3400175 3400304 3400406 3400507 3400608 3400811 3400895 3401318 3401521 3401503 3404565 3400785 3400911
DENE3400204EA 11190 DKK
DENE3400204 DENE3402732 DENE3400315 DENE3400102 DENE3400103 DENE3400115 DENE3400203 DENE3400305 DENE3400405 DENE3400501 DENE3400506 DENE3400516 DENE3400607 DENE3400613 DENE3400810 DENE3400813 DENE3401012 DENE3401214 DENE3400175 DENE3400304 DENE3400406 DENE3400507 DENE3400608 DENE3400811 DENE3400895 DENE3401318 DENE3401521 DENE3401503 DENE3404565 DENE3400785 DENE3400911
Tubing, PTFE
Slanger Overførselsslange
PTFE, white, translucent.
  • Excellent chemical resistance
  • Excellent thermal resistance, autoclavable
  • Free of extractable substances, physiologically neutral, biocompatible
This tubing is characterised by its non stick surface and excellent dielectric properties.
Temperature resistance -200 to +200 °C.