Tubing, rubber
Leverandør: DEUTSCH NEUMANN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3010204A 3010305 3010407 3010508 3010609 3010711 3010406 3010508A 3010609A 3010610A 3010710 3010711A 3010812 3010913B 3011014B 3011217B 3011824 3012533 3010408A 3011622 3012028 3011420 3010509 3011521
228-3331EA 45.7 DKK
228-3331 228-3335 228-3336 228-3344 228-3346 228-3356 228-3812 228-3813 228-3814 228-3815 228-3816 228-3817 228-3818 228-3819 228-3820 228-3821 228-3822 228-1544 228-1537 228-1542 228-1543 228-1540 228-1538 228-1541
Tubing, rubber
Slanger Overførselsslange
Natural rubber, red.
  • Excellent elasticity and flexibility
  • Wear resistant
  • Good chemical resistance to acids and alkalis
This tubing is light, flexible and durable. Its elasticity makes it easy to handle and provides a tight seal.
Temperature resistance: −10 to +50 °C
Hardness (Shore durometer A): 50
Density: 1.13