Gas inlet/air bleed tubes
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
05080001 05080002
201-2611EA 172 DKK
201-2611 612-2816
Gas inlet/air bleed tubes
Rør Gas indgangsrør
Duran® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Straight inlet tube with cone, that can be cut to required length or drawn as a capillary for an air/gas bleed.
Useful length: 200 mm
Certificeringer: Gevindadapter med DIN-gevind og standard kolbe med slibning.