Drying tubes with standard ground joints, U-shaped
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
05342013 05342015
201-9122EA 1140 DKK
201-9122 201-9120
Drying tubes with standard ground joints, U-shaped
Rør Tørrerør
DURAN®, borosilicate glass, clear. Replaceable NS stopcocks. Lateral tube connectors with 6 mm external diameter.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes