Test tubes with standard ground stoppers
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1628/06M 1628/10M 1628/18M 1628/22M
212-7432EA 1730 DKK
212-7432 212-7433 212-7399 212-7398
Test tubes with standard ground stoppers
Rør Reagensglas
Pyrex® borosilicate clear glass.
  • Graduated
  • Excellent chemical and thermal resistance
  • Interchangeable PE stopper (included)
Ground socket tubes with white enamel graduations to indicate capacity.
Certificeringer: ISO 4142, DIN 12217