Test tubes without rim
Leverandør: GLASWARENFABRIK KARL HECHT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
42775009 42775024 42775018 42775251 42775051 42775054 42775030 42775060 42775012 42775027 42775003 42775045 42775042 42775015 42775039 42775033 42775021
212-1700EA 108 DKK
212-1700 212-1704 HECH42775018 HECH42775251 HECH42775051 HECH42775054 HECH42775030 HECH42775060 HECH42775012 HECH42775027 212-0290 HECH42775045 HECH42775042 HECH42775015 HECH42775039 HECH42775033 HECH42775021
Test tubes without rim
Rør Reagensglas
AR-Glas® (soda-lime glass).
  • Heavy wall (212-1700 suitable as Durham-Tube)
  • Good chemical and physical properties
  • Can also be used as disposable test tubes