Tubes, rack packed, non-sterile
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
781525 781535 781500 781510 781520 781530 781540
211-3255EA 817 DKK
211-3255 211-3257 211-3252 211-3253 211-3254 211-3256 391-5701
Tubes, rack packed, non-sterile
Rør Mikrorør
PP tubes and PE caps, non-sterile.
  • Tubes and racks are autoclavable at 121 °C (2 bar) according to DIN EN 285 (not caps)
  • Designed for storing and freezing down to −80 °C and for transporting reagents and samples
  • Compatible with standard microplates
  • Suitable for working with multi-channel pipettors
Suitable for biological tests, such as PCR, cell uptake studies, RIA and EIA.
Tube o. Ø×h: 8,8×45 mm