VWR®, Strimler af PCR rør og hætter
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
732-0546EA 1910 DKK
732-0546 732-0550 732-0552 732-0553 732-0554 732-0545 211-0338 211-0339 211-0381 732-1506 732-1521 732-1517 732-1518 732-1519 732-1520 731-0367 732-3391 732-3484 732-3485 732-3499 732-3608 732-3609 732-3610
VWR®, Strimler af PCR rør og hætter
Rør PCR rør
These PCR tube strips and caps are made of PP resin and are designed to fit most popular brands of thermal cyclers.
  • Certified free from DNase, RNase and human DNA
  • Manufactured from polypropylene under strict quality control guidelines
Available with domed or flat caps.