Melting point and boiling point tubes and capillaries
Leverandør: BUCHI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
001759 001762 017808 019007
621-0003EA 2370 DKK
621-0003 621-0004 621-0005 621-0006
Melting point and boiling point tubes and capillaries
Rør Smeltepunktsrør
For an accurate determination of a melting point & boiling point, the quality of the used tubes and capillaries is decisive. The original Büchi glass components guarantee uniform wall thickness at the sides and ends of the tubes and capillaries.
  • For use with melting point apparatus B-540/545
  • Capillaries dimensions 80×1,55 mm (L×W)
  • Capillary wall thickness is 0,225 mm
  • Tube vial dimensions 92×24 mm (L×W)