Capillaries for melting point determination
Leverandør: MARIENFELD
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2930207 2930210 2930205 2940205 2930208 2940208 2930201 2930203 2930204 2940206 2940207 2940209 2940200 2940210 2940202
621-1967EA 1880 DKK
621-1967 621-1968 621-1920 621-1921 621-1930 621-1931 621-1964 621-1965 621-1966 621-1970 621-1971 621-1972 MARI2940200 621-1963 MARI2940202
Capillaries for melting point determination
Rør Smeltepunktsrør
These single use, glass capillary tubes can be used for storage of small amounts of liquids or for the determination of melting point.
  • With rectangular cut edges
  • Tolerance length: ± 1 mm
  • Tolerance diameter: ± 0,05 mm
Type A capillaries have two open ends
Type B capillaries have one closed end