Centrifugerør, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
216011004 216011107 216011407 216011707 216012403 216012609 216013605 216111705 242630901 216111405
525-1102EA 74.5 DKK
525-1102 525-1104 525-1106 525-1108 525-1110 525-1112 525-1114 525-1142 525-1122 525-1132
Centrifugerør, DURAN®
Rør Centrifugerør
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical and thermal resistance
  • Max. RCF: 4000×g
Robust universal centrifuge tubes for a wide variety of applications.
525-1110 and 525-1114 not in accordance with DIN.
Certificeringer: DIN 58970 (Part 2) only for round bottom tubes.