Centrifuge tubes
Leverandør: VITLAB
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
103897 103797
525-2250EA 760 DKK
525-2250 525-2251
Centrifuge tubes
Rør Centrifugerør
PFA.
  • Translucent
  • Withstand forces up to 10000×g
  • Also for autosampler racks
Bestillingsoplysninger: 525-2251 without screw cap, 525-2250 with calibrated ring-mark at 10 ml, thread GL25 and screw cap.
Advarsel: *The product(s) on this page are outside the scope of the EU Directive 2019/904