Laboratoriebakker, lav eller høj kant
Leverandør: Gilac PROFESSIONNEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
G154121 G154221 G154321 G154421 G154521 G110021 G155421 G113721 G155021
216-2881EA 80.4 DKK
216-2881 216-2882 216-2883 216-2884 216-2885 216-2864 216-2887 216-2865 216-2886
Laboratoriebakker, lav eller høj kant
Bakker/Tallerkener/Potter Laboratoriebakker/-skåle
HDPE (low profile) or LDPE (high profile), translucent.
  • Alimentary use
  • Thermal resistance −15 to +60 °C (HDPE), −40 to +90 °C (LDPE)
  • Lightweight and sturdy
Ideal for a wide range of uses.