Trays, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7120-0010 7120-0020
216-0329EA 488 DKK
216-0329 216-0330
Trays, Nalgene®
Bakker/Tallerkener/Potter Laboratoriebakker/-skåle
HDPE, semi-transparent.
  • Good chemical resistance
  • Heat-resistant from −100 to +120 °C
  • Tough, stackable
Universal trays for a wide range of applications. Ideal for soaking laboratory equipment or as an ice bath. Slightly tapered walls for better stackability. With handles for easy transportation.