Multipurpose tongs, stainless steel
Leverandør: HAMMACHER KARL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HWN310-20 HWN312-28
231-0279EA 922 DKK
231-0279 HAMMHWN312-28
Multipurpose tongs, stainless steel
Tænger
Stainless steel 18/10, anti-magnetic. Straight design for a wide range of laboratory, industrial and medical applications.
  • Acid-resistant
  • Excellent thermal resistance, autoclavable