VWR®, Præcisiontermometre, frio temp
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
620-0837EA 933 DKK
620-0837 620-0838 620-0839 620-0840 620-0841
VWR®, Præcisiontermometre, frio temp
Termometre Analoge termometre
Kan bruges i frysere, køleskabe, inkubatorer, ovne og blodbanker. Ved brug skal termometer og låg blot skrues på flasken.
  • Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
  • Thermometers contain organic liquid; featuring permanently fused markings and lead-free glass
  • PFA coating safely contains glass and liquid in case of breakage
  • Individual serial number
Insert the thermometer into the bottle and place near the samples for best results.
Division: 0,1 °C
Traceable to NIST/DKD
Certificeringer: Traceable to NIST.
Leveringsinformation: Supplied with a multi-language (English, French, German, Italian, Portuguese and Spanish) statement of accuracy and traceability plus VELCRO® and magnetic attachments.