VWR®, Termometre for varmeblokke og vandbade
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
620-0816EA 600 DKK
620-0816 620-0817 620-0818 620-0820 620-0821 620-0822 620-0887 620-0889
VWR®, Termometre for varmeblokke og vandbade
Termometre Analoge termometre
Contains red organic liquid against white back glass, except for 620-0891 which has yellow back glass.
  • For use in all standard heating blocks and incubators
  • PTFE coating keeps liquid and glass encapsulated in the event of breakage
  • Individually serialised
Accuracy: ±1,0 °C (620-0888: ±0,5 °C)