VWR®, Miljøvenlige termometre, easy read
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
620-0781EA 190 DKK
620-0781 620-0782 620-0784 620-0785 620-0786 620-0787 620-0788 620-0789
VWR®, Miljøvenlige termometre, easy read
Termometre Analoge termometre
These thermometers are easy to read quickly and accurately due to the high contrast black numbering against a yellow background. Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
  • Enviro safe liquid
  • Eliminates reading errors
  • Non toxic, non hazardous, biodegradable liquid
Traceable to NIST/DKD. Compliant with Envirokleen™.
Certificeringer: Traceable NIST. Envirokleen certified by ChemTel (an independentMore Product Information