Syringes, FORTUNA® OPTIMA®, record tip
Leverandør: Poulten & Graf
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7.20027 7.20033 7.20037 7.20041 7.20044
613-0491EA 2330 DKK
613-0491 613-0492 POUL7.20037 613-0493 POUL7.20044
Syringes, FORTUNA® OPTIMA®, record tip
Sprøjter Sprøjter, medicinske
Barrel made from Durobax® glass, metal tip and interchangeable piston, no needle, autoclavable.
  • Record tip, interchangeable piston
  • Graduations in brown
  • Autoclavable at +134 °C
Certificeringer: Conform to ISO 9001, medical directive 93/42/EEC.
Bestillingsoplysninger: Supplied in individual packaging.