Syringes, FORTUNA® OPTIMA®, luer lock tip
Leverandør: Poulten & Graf
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7.14021 7.14027 7.14029 7.14033 7.14037 7.14041 7.14044 7.14045 7.14051
612-0331EA 161 DKK
612-0331 612-0332 612-0333 612-0587 612-0335 612-0336 612-0337 612-0338 612-0339
Syringes, FORTUNA® OPTIMA®, luer lock tip
Sprøjter Sprøjter, medicinske
Barrel and piston in Durobax® glass with metal tip, no needle.
  • Luer lock tip
  • Graduations in brown
  • Autoclavable at +134 °C
Certificeringer: Conform to ISO 9001, medical directive 93/42/EEC.
Bestillingsoplysninger: Supplied individually packaged.