Precision gas syringes, FORTUNA®
Leverandør: Poulten & Graf
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2.00043 2.00046 2.00051 2.01246 2.01251 2.01651
201-0918EA 949 DKK
201-0918 201-0919 612-0397 201-0920 612-0398 201-0921
Precision gas syringes, FORTUNA®
Sprøjter Sprøjter, medicinske
Precision gas syringes made from DURAN®, borosilicate glass.
  • With or without capillary stopcock NS 12,5
  • Amber graduation, acid and alkali resistant
  • Interchangeable cylinders and pistons