Syringes for gas chromatography autosamplers, CTC PAL® GC-xt and Combi PAL-xt liquid mode
Leverandør: HAMILTON BONADUZ
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
202668 203043 203072 203074 203075 203076 203078 203080 203185 203189 203197 203205 203206 203209 203219 203225 203226 203363 203362 203361 203275
HAMI202668EA 1150 DKK
HAMI202668 549-1313 549-1310 549-0569 549-1319 549-1314 549-1315 549-1316 549-1307 549-1311 549-0570 549-1312 549-0572 549-0573 549-0574 HAMI203225 549-0575 549-1308 549-0525 549-1309 HAMI203275
Syringes for gas chromatography autosamplers, CTC PAL® GC-xt and Combi PAL-xt liquid mode
Sprøjter Sprøjter til kromatografi
These C-line syringes feature a direct attachment design of the needle to the barrel which eliminates contact between the sample solvent and adhesive, reducing carryover and ghost peaks. The inert plunger tip polymer provides enhanced solvent compatibility and longer lifetime.