Transport swabs, Transwab®
Leverandør: MEDICAL WIRE EQUIPMENT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MW170 MW171 MW172P MW173C MW173P MW172C
720-0138EA 837 DKK
720-0138 720-0139 720-0140 720-2201 720-2202 MEDWMW172C
Transport swabs, Transwab®
Podepinde Podepinde til transport
Each transport tube contains Amies medium preferred because the inorganic buffer ensures maintenance without overgrowth.
  • Excellent recovery of both aerobic and anaerobic bacteria
  • High release of micro-organisms onto plates or into broth
  • Safety cap includes double seal, inside and outside tube
  • Colour coded caps for ease of selection
  • Pre-labelled transport tube
Certificeringer: MDD, IVD, and CLSI M40.
Emballering: Easy peel pack.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904