Hollow-blown glass stoppers, B-length, Quickfit®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SB10 SB19 SB24 SB45 SB14 SB34 SB29 SB12
201-0528EA 492 DKK
201-0528 201-8674 201-8675 201-0531 201-8672 201-2376 201-0530 BIBBSB12
Hollow-blown glass stoppers, B-length, Quickfit®
Propper Glas
These glass stoppers are ideally suited for use with Quickfit® jointed glassware and other glassware having standard ground glass sockets.
  • Chemical resistance
  • High temperature resistance
  • Hollow-blown construction to reduce weight
  • Hexagonal head prevents the stopper from rolling and provides a good grip
All Quickfit® stoppers conform to the standard tapers as outlined in ISO 383.
ISO 383