Rods, glass
Leverandør: FISCHER LABORTECHNIK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2012003 2012004 2012005 2012006 2012007 2012008
201-2003EA 298 DKK
201-2003 201-2004 201-2005 201-2006 201-2007 201-2008
Rods, glass
Stænger
DURAN®, borosiicate glass.
  • Appropriately cut to 500 mm in length (cut ends)
  • Excellent chemical and thermal resistance
  • Permissible diameter tolerance of ±0.4 mm